Product Details

SNP ID
rs73446054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73736048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGGTCAGTGGAATAGGACTTCC[C/T]GGAAGCAGAGAGATCTGATGGAACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C15orf59 PubMed Links

Gene Details

Gene
C15orf59
Gene Name
chromosome 15 open reading frame 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039614.2 4979 UTR 3 NP_001034703.1
NM_001303254.1 4979 UTR 3 NP_001290183.1
XM_005254369.4 4979 UTR 3 XP_005254426.1

View Full Product Details