Product Details

SNP ID
rs74026858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:87577931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAATTAAATCTGATTTCTCTGA[A/G]CCTCAGTTTCCTAATCTGAGAAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00052 PubMed Links

Gene Details

Gene
LINC00052
Gene Name
long intergenic non-protein coding RNA 52
There are no transcripts associated with this gene.

View Full Product Details