Product Details

SNP ID
rs72766457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:8623611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCCATCACTTTCAATGACACC[A/G]TGAAATTCTGCATTGATTTGCCAGA
Phenotype
MIM: 615261
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
METTL22 PubMed Links

Gene Details

Gene
METTL22
Gene Name
methyltransferase like 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024109.3 Intron NP_077014.3
XM_005255570.3 Intron XP_005255627.1
XM_005255571.2 Intron XP_005255628.1
XM_011522657.2 Intron XP_011520959.1
XM_017023659.1 Intron XP_016879148.1
XM_017023660.1 Intron XP_016879149.1
XM_017023661.1 Intron XP_016879150.1
XM_017023662.1 Intron XP_016879151.1

View Full Product Details