Product Details

SNP ID
rs73301793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21001787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTAGTTTCTAATTTCTCAGTGGA[C/T]AAATGGACAAACCATCTCTGTTTGA
Phenotype
MIM: 612116
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105369205 PubMed Links

Gene Details

Gene
LOC105369205
Gene Name
uncharacterized LOC105369205
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011524128.2 3026 Intron XP_011522430.1
XM_011524129.2 3026 Intron XP_011522431.1
XM_011524131.2 3026 Intron XP_011522433.1
XM_017025507.1 3026 Intron XP_016880996.1
XM_017025508.1 3026 Intron XP_016880997.1
XM_017025509.1 3026 Intron XP_016880998.1
Gene
USP22
Gene Name
ubiquitin specific peptidase 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015276.1 3026 UTR 3 NP_056091.1
XM_005256575.2 3026 UTR 3 XP_005256632.1

View Full Product Details