Product Details

SNP ID
rs72843524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60223610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTATAAAAAAAAAAGAGGATAGA[A/T]TCTCTTATTTTTAGTTATTATACAT
Phenotype
MIM: 607740
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SCARNA20 PubMed Links

Gene Details

Gene
SCARNA20
Gene Name
small Cajal body-specific RNA 20
There are no transcripts associated with this gene.

Gene
USP32
Gene Name
ubiquitin specific peptidase 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032582.3 Intron NP_115971.2
XM_011525371.1 Intron XP_011523673.1
XM_011525372.1 Intron XP_011523674.1
XM_011525373.1 Intron XP_011523675.1
XM_011525374.1 Intron XP_011523676.1
XM_011525375.1 Intron XP_011523677.1
XM_011525376.1 Intron XP_011523678.1
XM_011525378.1 Intron XP_011523680.1
XM_011525379.2 Intron XP_011523681.1
XM_017025233.1 Intron XP_016880722.1
XM_017025234.1 Intron XP_016880723.1

View Full Product Details