Product Details

SNP ID
rs72868358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:5893055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTATCATGTGTCAGATTTCATCA[A/G]GGAATTTGAGAAGTCCCCTGGAATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM200C PubMed Links

Gene Details

Gene
TMEM200C
Gene Name
transmembrane protein 200C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080209.1 5050 Intron NP_001073678.1
XM_005258130.4 5050 Intron XP_005258187.1
XM_011525729.2 5050 UTR 5 XP_011524031.1

View Full Product Details