Product Details

SNP ID
rs73943346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:655442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGATCTTCCTCGTGCTCGGAAAC[A/G]TGTAGGGAAACGTGTAGGGACAGTT
Phenotype
MIM: 616990 MIM: 188350
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLUL1 PubMed Links

Gene Details

Gene
CLUL1
Gene Name
clusterin like 1
There are no transcripts associated with this gene.

Gene
TYMS
Gene Name
thymidylate synthetase
There are no transcripts associated with this gene.

Gene
TYMSOS
Gene Name
TYMS opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012716.2 Intron NP_001012734.2

View Full Product Details