Product Details

SNP ID
rs73428511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35334763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGGAATTTAAAAATCACTACA[C/T]AGTATTTAAAATCAATTTGTCTAGT
Phenotype
MIM: 194534
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF24 PubMed Links

Gene Details

Gene
ZNF24
Gene Name
zinc finger protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308123.1 3754 Intron NP_001295052.1
NM_006965.3 3754 UTR 3 NP_008896.2
XM_005258341.3 3754 UTR 3 XP_005258398.1

View Full Product Details