Product Details

SNP ID
rs73044923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38396502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTGTTGAAATGAACAACCTAGA[A/C]CCCGGTCACGCCTGGCACCATGTCC
Phenotype
MIM: 609966 MIM: 609293
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM98C PubMed Links

Gene Details

Gene
FAM98C
Gene Name
family with sequence similarity 98 member C
There are no transcripts associated with this gene.

Gene
GGN
Gene Name
gametogenetin
There are no transcripts associated with this gene.

Gene
SPRED3
Gene Name
sprouty related EVH1 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042522.2 1693 UTR 3 NP_001035987.1
XM_006723219.3 1693 UTR 3 XP_006723282.1
XM_011526978.2 1693 UTR 3 XP_011525280.1

View Full Product Details