Product Details

SNP ID
rs73593647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34924635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCAGTGACCACAGCCAGTTCAGG[A/G]ATCACAGGATATTGCAAATTTGTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF30 PubMed Links

Gene Details

Gene
ZNF30
Gene Name
zinc finger protein 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099437.1 Intron NP_001092907.1
NM_001099438.1 Intron NP_001092908.1
NM_194325.2 Intron NP_919306.2
XM_011527443.2 Intron XP_011525745.1
XM_011527444.1 Intron XP_011525746.1
XM_017027423.1 Intron XP_016882912.1
XM_017027424.1 Intron XP_016882913.1
XM_017027425.1 Intron XP_016882914.1
XM_017027426.1 Intron XP_016882915.1
XM_017027427.1 Intron XP_016882916.1
XM_017027428.1 Intron XP_016882917.1
XM_017027429.1 Intron XP_016882918.1
Gene
ZNF30-AS1
Gene Name
ZNF30 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details