Product Details

SNP ID
rs73304831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45648889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGTTGTCCACTTCTGAGTAGGT[G/T]AGGTATTGGACAAAAAAAGGTGGAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WFDC11 PubMed Links

Gene Details

Gene
WFDC11
Gene Name
WAP four-disulfide core domain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147197.2 Intron NP_671730.1

View Full Product Details