Product Details

SNP ID
rs73911119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45835018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCTCCTGGCGCGGCAGCTGCAG[A/G]ATTTCTGGAAGAAGTCCCGGAACAC
Phenotype
MIM: 608123 MIM: 191039
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACOT8 PubMed Links

Gene Details

Gene
ACOT8
Gene Name
acyl-CoA thioesterase 8
There are no transcripts associated with this gene.

Gene
SNX21
Gene Name
sorting nexin family member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042632.2 438 Missense Mutation AAT,GAT N117D NP_001036097.1
NM_001042633.2 438 Missense Mutation AAT,GAT N117D NP_001036098.1
NM_033421.3 438 Missense Mutation AAT,GAT N117D NP_219489.1
NM_152897.2 438 Missense Mutation AAT,GAT N117D NP_690857.1
XM_005260608.4 438 Intron XP_005260665.1
XM_011529095.2 438 Intron XP_011527397.1
XM_011529096.2 438 Missense Mutation AAT,GAT N117D XP_011527398.1
XM_011529098.2 438 Missense Mutation AAT,GAT N117D XP_011527400.1
XM_011529099.2 438 Missense Mutation AAT,GAT N117D XP_011527401.1
XM_017028120.1 438 Missense Mutation AAT,GAT N117D XP_016883609.1
Gene
TNNC2
Gene Name
troponin C2, fast skeletal type
There are no transcripts associated with this gene.

View Full Product Details