Product Details

SNP ID
rs73902900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:22560829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCATGGACATTTTAGAGGCAGTC[G/T]TAAGTCTGCTGTGGGCACAGGCATG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00261 PubMed Links

Gene Details

Gene
LINC00261
Gene Name
long intergenic non-protein coding RNA 261
There are no transcripts associated with this gene.

View Full Product Details