Product Details

SNP ID
rs67976848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:45922298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTCTAGGGGATGGGCTGACACA[C/G]AGTACGGAGTTAGGACTGGCTATGT
Phenotype
MIM: 601967
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
WNT7B PubMed Links

Gene Details

Gene
WNT7B
Gene Name
Wnt family member 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058238.2 1982 UTR 3 NP_478679.1
XM_011530366.1 1982 UTR 3 XP_011528668.1

View Full Product Details