Product Details

SNP ID
rs73214921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119581966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCATTGGACGCATATCAGAATTA[A/C]TGTGGAACATTTTAAAAACACAGAT
Phenotype
MIM: 300440 MIM: 312180
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NKRF PubMed Links

Gene Details

Gene
NKRF
Gene Name
NFKB repressing factor
There are no transcripts associated with this gene.

Gene
UBE2A
Gene Name
ubiquitin conjugating enzyme E2 A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282161.1 Intron NP_001269090.1
NM_003336.3 Intron NP_003327.2
NM_181762.2 Intron NP_861427.1

View Full Product Details