Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001300823.1 | 4444 | UTR 3 | NP_001287752.1 | ||
NM_001300824.1 | 4444 | UTR 3 | NP_001287753.1 | ||
NM_018699.3 | 4444 | UTR 3 | NP_061169.2 | ||
XM_005262708.3 | 4444 | Intron | XP_005262765.1 | ||
XM_011531562.2 | 4444 | UTR 3 | XP_011529864.1 | ||
XM_011531563.2 | 4444 | UTR 3 | XP_011529865.1 | ||
XM_011531564.2 | 4444 | UTR 3 | XP_011529866.1 | ||
XM_011531565.2 | 4444 | Intron | XP_011529867.1 | ||
XM_011531566.2 | 4444 | Intron | XP_011529868.1 | ||
XM_011531567.2 | 4444 | Intron | XP_011529869.1 | ||
XM_011531568.2 | 4444 | Intron | XP_011529870.1 | ||
XM_011531569.2 | 4444 | Intron | XP_011529871.1 | ||
XM_011531570.2 | 4444 | UTR 3 | XP_011529872.1 | ||
XM_017007668.1 | 4444 | UTR 3 | XP_016863157.1 | ||
XM_017007669.1 | 4444 | Intron | XP_016863158.1 | ||
XM_017007670.1 | 4444 | Intron | XP_016863159.1 | ||
XM_017007671.1 | 4444 | UTR 3 | XP_016863160.1 |