Product Details

SNP ID
rs72994458
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:114719698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTGAACTTCTTTCACATGGAA[A/G]AAGAGCTTCTTGCTATCTCCTACTT
Phenotype
MIM: 191510 MIM: 164790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSDE1 PubMed Links

Gene Details

Gene
CSDE1
Gene Name
cold shock domain containing E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007553.2 2223 Silent Mutation TTC,TTT F699F NP_001007554.1
NM_001130523.2 2223 Silent Mutation TTC,TTT F714F NP_001123995.1
NM_001242891.1 2223 Silent Mutation TTC,TTT F745F NP_001229820.1
NM_001242892.1 2223 Silent Mutation TTC,TTT F699F NP_001229821.1
NM_001242893.1 2223 Silent Mutation TTC,TTT F668F NP_001229822.1
NM_007158.5 2223 Silent Mutation TTC,TTT F668F NP_009089.4
Gene
NRAS
Gene Name
neuroblastoma RAS viral oncogene homolog
There are no transcripts associated with this gene.

View Full Product Details