Product Details

SNP ID
rs72470572
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:220786988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGCGCTGGCGCGCTTTGTCCTCC[A/T]CGCGCAATCCCGGCCCGGGTGGCTC
Phenotype
MIM: 614126 MIM: 614127
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MARC1 PubMed Links

Gene Details

Gene
MARC1
Gene Name
mitochondrial amidoxime reducing component 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022746.3 292 Missense Mutation CAC,CTC H15L NP_073583.3
XM_011509900.2 292 Missense Mutation CAC,CTC H15L XP_011508202.1
XM_011509903.2 292 Intron XP_011508205.1
XM_011509904.2 292 Intron XP_011508206.1
XM_017002096.1 292 Intron XP_016857585.1
XM_017002097.1 292 Intron XP_016857586.1
Gene
MARC2
Gene Name
mitochondrial amidoxime reducing component 2
There are no transcripts associated with this gene.

View Full Product Details