Product Details

SNP ID
rs73953278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127187137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAGATTTACAAGTGTCCCAGGA[C/T]CTGGGAGGCCAGTCTATAACAAGAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP27C1 PubMed Links

Gene Details

Gene
CYP27C1
Gene Name
cytochrome P450 family 27 subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001665.3 1384 UTR 3 NP_001001665.3
XM_017003960.1 1384 UTR 3 XP_016859449.1
XM_017003961.1 1384 Intron XP_016859450.1

View Full Product Details