Product Details

SNP ID
rs73961768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:143073785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTAGGCTCCATAAAAACTCTAG[A/C]TTTTCTCTTGTTTGCCCTCTCTTGT
Phenotype
MIM: 610578
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP15 PubMed Links

Gene Details

Gene
ARHGAP15
Gene Name
Rho GTPase activating protein 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018460.3 Intron NP_060930.3
XM_011511479.2 Intron XP_011509781.1
XM_011511481.2 Intron XP_011509783.1
XM_011511482.2 Intron XP_011509784.1
XM_011511483.1 Intron XP_011509785.1
XM_011511484.1 Intron XP_011509786.1
XM_017004499.1 Intron XP_016859988.1
XM_017004500.1 Intron XP_016859989.1
XM_017004501.1 Intron XP_016859990.1

View Full Product Details