Product Details

SNP ID
rs73829668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44594420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAGCGAATCCACACGGGAGAGA[A/G]ACCCTATAAGTGTAAGGAGTGTGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF660 PubMed Links

Gene Details

Gene
ZNF660
Gene Name
zinc finger protein 660
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173658.2 560 Missense Mutation AAA,AGA K76R NP_775929.2
XM_005265090.4 560 Missense Mutation AAA,AGA K76R XP_005265147.1
XM_006713117.3 560 Missense Mutation AAA,AGA K76R XP_006713180.1
XM_011533622.2 560 Missense Mutation AAA,AGA K76R XP_011531924.1

View Full Product Details