Product Details

SNP ID
rs73074478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44629125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCAAGGATTAAGGAGACCTGGA[A/C]TGGAGAGGAGCCTTTTTCAAAAAAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF197 PubMed Links

Gene Details

Gene
ZNF197
Gene Name
zinc finger protein 197
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005264783.4 160 UTR 5 XP_005264840.1
XM_017005495.1 160 UTR 5 XP_016860984.1
Gene
ZNF197-AS1
Gene Name
ZNF197 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details