Product Details

SNP ID
rs116232180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:47562596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCCGCTTCCTCTCTTCTTGCTC[C/T]GCTTTAGGTTAAATTATTCTGAAGA
Phenotype
MIM: 606775
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSPG5 PubMed Links

Gene Details

Gene
CSPG5
Gene Name
chondroitin sulfate proteoglycan 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206942.1 1959 UTR 3 NP_001193871.1
NM_001206943.1 1959 UTR 3 NP_001193872.1
NM_001206944.1 1959 UTR 3 NP_001193873.1
NM_001206945.1 1959 UTR 3 NP_001193874.1
NM_006574.3 1959 UTR 3 NP_006565.2

View Full Product Details