Product Details

SNP ID
rs73074850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49562187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCTTCTACCTCCTACTGTGTCA[C/T]CTACACTTTTTTGTGTATGGGAGTG
Phenotype
MIM: 604020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BSN PubMed Links

Gene Details

Gene
BSN
Gene Name
bassoon presynaptic cytomatrix protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003458.3 Intron NP_003449.2
Gene
BSN-AS2
Gene Name
BSN antisense RNA 2 (head to head)
There are no transcripts associated with this gene.

View Full Product Details