Product Details

SNP ID
rs73182407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56819914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGCGTGAGTCACTGCGCCCAGC[C/T]GCAAAATGGTTAGGATTTGATTACT
Phenotype
MIM: 605753
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPINK2 PubMed Links

Gene Details

Gene
SPINK2
Gene Name
serine peptidase inhibitor, Kazal type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271718.1 Intron NP_001258647.1
NM_001271719.1 Intron NP_001258648.1
NM_001271720.1 Intron NP_001258649.1
NM_001271721.1 Intron NP_001258650.1
NM_001271722.1 Intron NP_001258651.1
NM_021114.3 Intron NP_066937.1
XM_011534405.1 Intron XP_011532707.1
XM_011534406.2 Intron XP_011532708.1

View Full Product Details