Product Details

SNP ID
rs73572789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:28431612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCCCCAGAAAGGTCCGTCTTCA[C/T]AGCACATAGACTTAACAGCAAGAAG
Phenotype
MIM: 608519
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO16 PubMed Links

Gene Details

Gene
FBXO16
Gene Name
F-box protein 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258211.1 Intron NP_001245140.1
NM_172366.3 Intron NP_758954.1

View Full Product Details