Product Details
- SNP ID
-
rs73476338
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:83663434 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAACACAAGCTACCGCAGGAGGTG[A/G]TGTCATGGGTGGAGTTAAGGTAGTT
- Phenotype
-
MIM: 605046
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UBQLN1
PubMed Links
Gene Details
- Gene
- UBQLN1
- Gene Name
- ubiquilin 1
View Full Product Details