Product Details

SNP ID
rs73289738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:50109702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGTAAGATGATTAAAGGCTTCA[A/C]GATGGAAATTTCTATAACAAAAAAA
Phenotype
MIM: 601247 MIM: 615260
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC01599 PubMed Links

Gene Details

Gene
LINC01599
Gene Name
long intergenic non-protein coding RNA 1599
There are no transcripts associated with this gene.

Gene
SOS2
Gene Name
SOS Ras/Rho guanine nucleotide exchange factor 2
There are no transcripts associated with this gene.

Gene
VCPKMT
Gene Name
valosin containing protein lysine methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040662.1 Intron NP_001035752.1
NM_024558.2 Intron NP_078834.2
XM_005268060.3 Intron XP_005268117.1
XM_017021640.1 Intron XP_016877129.1
XM_017021641.1 Intron XP_016877130.1
XM_017021642.1 Intron XP_016877131.1

View Full Product Details