Product Details

SNP ID
rs67748933
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:58437730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAAAAAAAAAAAAAAAAAAAA[A/G]GCCAAGTTCACAAAAGGGATCCAAG
Phenotype
MIM: 610178
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KIAA0586 PubMed Links

Gene Details

Gene
KIAA0586
Gene Name
KIAA0586
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244189.1 Intron NP_001231118.1
NM_001244190.1 Intron NP_001231119.1
NM_001244191.1 Intron NP_001231120.1
NM_001244192.1 Intron NP_001231121.1
NM_001244193.1 Intron NP_001231122.1

View Full Product Details