Product Details

SNP ID
rs73329641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:88467000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTGGGAAAAGCCCCAAGGAAAC[A/G]GAGTAAGTCACTGGCGGTCACAGGA
Phenotype
MIM: 603271 MIM: 609868
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTPN21 PubMed Links

Gene Details

Gene
PTPN21
Gene Name
protein tyrosine phosphatase, non-receptor type 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007039.3 4987 UTR 3 NP_008970.2
XM_005267287.2 4987 UTR 3 XP_005267344.1
XM_006720011.3 4987 UTR 3 XP_006720074.1
XM_011536367.2 4987 UTR 3 XP_011534669.1
XM_011536368.2 4987 UTR 3 XP_011534670.1
XM_011536369.2 4987 UTR 3 XP_011534671.1
XM_017020938.1 4987 UTR 3 XP_016876427.1
XM_017020939.1 4987 UTR 3 XP_016876428.1
Gene
SPATA7
Gene Name
spermatogenesis associated 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040428.3 4987 Intron NP_001035518.1
NM_018418.4 4987 Intron NP_060888.2
XM_005267851.1 4987 Intron XP_005267908.1
XM_005267852.1 4987 Intron XP_005267909.1
XM_005267854.1 4987 Intron XP_005267911.1
XM_005267855.1 4987 Intron XP_005267912.1
XM_006720204.1 4987 Intron XP_006720267.1
XM_006720205.1 4987 Intron XP_006720268.1
XM_011536951.1 4987 Intron XP_011535253.1
XM_011536952.1 4987 Intron XP_011535254.1
XM_011536953.1 4987 Intron XP_011535255.1
XM_017021452.1 4987 Intron XP_016876941.1
XM_017021453.1 4987 Intron XP_016876942.1
XM_017021454.1 4987 Intron XP_016876943.1
XM_017021455.1 4987 Intron XP_016876944.1
XM_017021456.1 4987 Intron XP_016876945.1
XM_017021457.1 4987 Intron XP_016876946.1

View Full Product Details