Product Details

SNP ID
rs73345164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:92938310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATACAGACCCCAGGGACATTAG[C/T]GAAGCCATTCAGCAGTTGTGGCCAG
Phenotype
MIM: 118910 MIM: 601838
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHGA PubMed Links

Gene Details

Gene
CHGA
Gene Name
chromogranin A
There are no transcripts associated with this gene.

Gene
ITPK1
Gene Name
inositol-tetrakisphosphate 1-kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017021262.1 Intron XP_016876751.1
XM_017021263.1 Intron XP_016876752.1
XM_017021264.1 Intron XP_016876753.1

View Full Product Details