Product Details

SNP ID
rs74074511
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:92939815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTAAACGTCTTTTAAGGACTGG[A/G]AGTATGACATAACAAACTTGAGCAA
Phenotype
MIM: 118910 MIM: 601838
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CHGA PubMed Links

Gene Details

Gene
CHGA
Gene Name
chromogranin A
There are no transcripts associated with this gene.

Gene
ITPK1
Gene Name
inositol-tetrakisphosphate 1-kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017021262.1 2990 UTR 3 XP_016876751.1
XM_017021263.1 2990 UTR 3 XP_016876752.1
XM_017021264.1 2990 UTR 3 XP_016876753.1

View Full Product Details