Product Details

SNP ID
rs73258122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32763834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTTCCACCCTCCCCCGAAGCTG[C/G]GAAGTGTTTGAATTAAAGGGAATTA
Phenotype
MIM: 602900
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNMT3B PubMed Links

Gene Details

Gene
DNMT3B
Gene Name
DNA methyltransferase 3 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207055.1 Intron NP_001193984.1
NM_001207056.1 Intron NP_001193985.1
NM_006892.3 Intron NP_008823.1
NM_175848.1 Intron NP_787044.1
NM_175849.1 Intron NP_787045.1
NM_175850.2 Intron NP_787046.1
XM_011528653.2 Intron XP_011526955.1
XM_011528654.2 Intron XP_011526956.1

View Full Product Details