Product Details

SNP ID
rs73331765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:98384771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGGTCAGCCTGGTGGAGGCAGC[A/G]GCTGTGTGCTCACCTGTGCATGGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PYROXD2 PubMed Links

Gene Details

Gene
PYROXD2
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032709.2 Intron NP_116098.2
XM_011540293.2 Intron XP_011538595.1
XM_011540301.2 Intron XP_011538603.1
XM_017016835.1 Intron XP_016872324.1
XM_017016836.1 Intron XP_016872325.1
XM_017016837.1 Intron XP_016872326.1
XM_017016838.1 Intron XP_016872327.1
XM_017016839.1 Intron XP_016872328.1
XM_017016840.1 Intron XP_016872329.1
XM_017016841.1 Intron XP_016872330.1

View Full Product Details