Product Details

SNP ID
hCV99322841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:46462519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAGTGACGATGAAGACCATCAC[G/A]TCCACGGAATCCTGGCAATGTTCAG
Phenotype
MIM: 601790
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
LINC00842 PubMed Links

Gene Details

Gene
LINC00842
Gene Name
long intergenic non-protein coding RNA 842
There are no transcripts associated with this gene.

Gene
LOC107984026
Gene Name
arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4-like
There are no transcripts associated with this gene.

Gene
NPY4R
Gene Name
neuropeptide Y receptor Y4
There are no transcripts associated with this gene.

View Full Product Details