Product Details

SNP ID
rs72899858
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:52406436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCCTGATATGTTGGTAGGCACTG[G/T]ACGCTCAGGGGTGAAAATCAAACAA
Phenotype
MIM: 601902 MIM: 617031
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ORC1 PubMed Links

Gene Details

Gene
ORC1
Gene Name
origin recognition complex subunit 1
There are no transcripts associated with this gene.

Gene
PRPF38A
Gene Name
pre-mRNA processing factor 38A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032864.3 Intron NP_116253.2
XM_011542315.2 Intron XP_011540617.1

View Full Product Details