Product Details

SNP ID
rs71637818
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:53767838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGATTTCTCCCATTAAAGTATAG[C/T]TTTCAAAGCAACCACCACGACAAAG
Phenotype
MIM: 610115
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDC1 PubMed Links

Gene Details

Gene
NDC1
Gene Name
NDC1 transmembrane nucleoporin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168551.1 2448 UTR 3 NP_001162023.1
NM_018087.4 2448 UTR 3 NP_060557.3
XM_011541766.2 2448 UTR 3 XP_011540068.1

View Full Product Details