Product Details

SNP ID
rs72679734
Assay Type
Functionally Tested
NCBI dbSNP Submissions
23
Location
Chr.1:62201295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAATTGTTATCAGAAAAATTTGT[C/T]CTTATGTGCCTGTAGAAGTTTACCC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
L1TD1 PubMed Links

Gene Details

Gene
L1TD1
Gene Name
LINE1 type transposase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details