Product Details

SNP ID
rs72974579
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:96728756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGAATCTGTAGAGCAAATTTGG[A/G]GAAAATTGGCATCATGACTACAATT
Phenotype
MIM: 608449
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTBP2 PubMed Links

Gene Details

Gene
PTBP2
Gene Name
polypyrimidine tract binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300985.1 Intron NP_001287914.1
NM_001300986.1 Intron NP_001287915.1
NM_001300987.1 Intron NP_001287916.1
NM_001300988.1 Intron NP_001287917.1
NM_001300989.1 Intron NP_001287918.1
NM_001300990.1 Intron NP_001287919.1
NM_021190.3 Intron NP_067013.1
XM_011541874.2 Intron XP_011540176.1
XM_011541875.2 Intron XP_011540177.1
XM_011541876.2 Intron XP_011540178.1
XM_017001951.1 Intron XP_016857440.1

View Full Product Details