Product Details
- SNP ID
-
rs74102304
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
35
- Location
-
Chr.1:99993576 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACAATGTTCGCCAACCTAAAATAC[A/G]TTTCCCTGGGAATTTTGGTCTTTCA
- Phenotype
-
MIM: 605632
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLC35A3
PubMed Links
Gene Details
- Gene
- SLC35A3
- Gene Name
- solute carrier family 35 member A3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001271684.1 |
836 |
Missense Mutation |
ATT,GTT |
I8V |
NP_001258613.1 |
NM_001271685.1 |
836 |
Missense Mutation |
ATT,GTT |
I50V |
NP_001258614.1 |
NM_012243.2 |
836 |
Missense Mutation |
ATT,GTT |
I8V |
NP_036375.1 |
XM_005270691.4 |
836 |
Missense Mutation |
ATT,GTT |
I8V |
XP_005270748.1 |
XM_011541135.2 |
836 |
Missense Mutation |
ATT,GTT |
I50V |
XP_011539437.1 |
XM_011541136.2 |
836 |
Missense Mutation |
ATT,GTT |
I8V |
XP_011539438.1 |
XM_011541137.2 |
836 |
Missense Mutation |
ATT,GTT |
I50V |
XP_011539439.1 |
XM_011541138.2 |
836 |
Missense Mutation |
ATT,GTT |
I50V |
XP_011539440.1 |
XM_017000869.1 |
836 |
Missense Mutation |
ATT,GTT |
I8V |
XP_016856358.1 |
XM_017000870.1 |
836 |
Missense Mutation |
ATT,GTT |
I8V |
XP_016856359.1 |
XM_017000871.1 |
836 |
Missense Mutation |
ATT,GTT |
I8V |
XP_016856360.1 |
XM_017000872.1 |
836 |
UTR 5 |
|
|
XP_016856361.1 |
View Full Product Details