Product Details

SNP ID
rs72984045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102581821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACGCTCAGCATGATTCCTGGCAC[A/T]CAGTAAGCCAGCCATCAAGAAATAG
Phenotype
MIM: 604629
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MMP20 PubMed Links

Gene Details

Gene
MMP20
Gene Name
matrix metallopeptidase 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004771.3 Intron NP_004762.2

View Full Product Details