Product Details

SNP ID
rs73494060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1088523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTCTACCAAGCATCCCTCTTTC[C/T]GCTCCCACCTGGCACTGGCAGCGTC
Phenotype
MIM: 614147 MIM: 182455
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1QTNF8 PubMed Links

Gene Details

Gene
C1QTNF8
Gene Name
C1q and tumor necrosis factor related protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207419.3 3109 UTR 3 NP_997302.2
XM_011522483.2 3109 Intron XP_011520785.1
Gene
SSTR5
Gene Name
somatostatin receptor 5
There are no transcripts associated with this gene.

Gene
SSTR5-AS1
Gene Name
SSTR5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details