Product Details

SNP ID
rs72774395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15100098 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACTTTGTTTCTGGATTATATTG[C/T]GTGAGATTATGCTTATCCAGACTAC
Phenotype
MIM: 603801 MIM: 606116
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf111 PubMed Links

Gene Details

Gene
C10orf111
Gene Name
chromosome 10 open reading frame 111
There are no transcripts associated with this gene.

Gene
NMT2
Gene Name
N-myristoyltransferase 2
There are no transcripts associated with this gene.

Gene
RPP38
Gene Name
ribonuclease P/MRP subunit p38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097590.2 Intron NP_001091059.1
NM_001265601.1 Intron NP_001252530.1
NM_006414.4 Intron NP_006405.2
NM_183005.4 Intron NP_892117.1
XM_006717363.1 Intron XP_006717426.1
XM_006717364.3 Intron XP_006717427.1
XM_011519293.1 Intron XP_011517595.1
XM_017015479.1 Intron XP_016870968.1
XM_017015480.1 Intron XP_016870969.1
XM_017015481.1 Intron XP_016870970.1

View Full Product Details