Product Details

SNP ID
rs77651834
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:1242886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGCGTGAAGACCCTGCTGTTGCT[C/T]TCCAGGCCTGAGACGGCCTCCAGGC
Phenotype
MIM: 615291 MIM: 616593
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B3GALT6 PubMed Links

Gene Details

Gene
B3GALT6
Gene Name
beta-1,3-galactosyltransferase 6
There are no transcripts associated with this gene.

Gene
FAM132A
Gene Name
family with sequence similarity 132 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014980.2 885 Silent Mutation GAA,GAG E253E NP_001014980.1
XM_011541435.2 885 Missense Mutation AAA,AGA K258R XP_011539737.1
XM_011541436.2 885 Missense Mutation AAA,AGA K252R XP_011539738.1
XM_011541437.1 885 Missense Mutation AAA,AGA K186R XP_011539739.1

View Full Product Details