Product Details

SNP ID
rs75007822
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:160487140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTAATTGTGGAGTAAATTGTGAT[A/G]GTATCATTTTCTCTAGGTGTCCAGA
Phenotype
MIM: 606446
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLAMF6 PubMed Links

Gene Details

Gene
SLAMF6
Gene Name
SLAM family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184714.1 985 Silent Mutation ACC,ACT T305T NP_001171643.1
NM_001184715.1 985 Silent Mutation ACC,ACT T255T NP_001171644.1
NM_001184716.1 985 Silent Mutation ACC,ACT T194T NP_001171645.1
NM_052931.4 985 Silent Mutation ACC,ACT T304T NP_443163.1
XM_017000215.1 985 Silent Mutation ACC,ACT T305T XP_016855704.1
XM_017000216.1 985 Silent Mutation ACC,ACT T256T XP_016855705.1
XM_017000217.1 985 Silent Mutation ACC,ACT T193T XP_016855706.1

View Full Product Details