Product Details

SNP ID
rs75708071
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:163070674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTCACTTAACTTGCATATGAAA[C/T]GAACCGTACAAAAATACAAGAGTGT
Phenotype
MIM: 602516
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RGS4 PubMed Links

Gene Details

Gene
RGS4
Gene Name
regulator of G-protein signaling 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102445.2 Intron NP_001095915.1
NM_001113380.1 Intron NP_001106851.1
NM_001113381.1 Intron NP_001106852.1
NM_005613.5 Intron NP_005604.1

View Full Product Details