Product Details

SNP ID
rs78902800
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:171138202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGATGTTCTCTCCAGACTCTGCT[A/T]CAGGCAATCATGAGCAAGAGGGTTG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FMO6P PubMed Links

Gene Details

Gene
FMO6P
Gene Name
flavin containing monooxygenase 6 pseudogene
There are no transcripts associated with this gene.

View Full Product Details