Product Details

SNP ID
rs78077552
Assay Type
Functionally Tested
NCBI dbSNP Submissions
13
Location
Chr.1:150928359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTATGGTCAAAAATCTGACCTTA[A/C]AATCAGCCGTAAATATAATTTTCAT
Phenotype
MIM: 604396
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
SETDB1 PubMed Links

Gene Details

Gene
SETDB1
Gene Name
SET domain bifurcated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145415.1 Intron NP_001138887.1
NM_001243491.1 Intron NP_001230420.1
NM_012432.3 Intron NP_036564.3
XM_005245641.3 Intron XP_005245698.1
XM_017002953.1 Intron XP_016858442.1
XM_017002954.1 Intron XP_016858443.1
XM_017002955.1 Intron XP_016858444.1

View Full Product Details