Product Details

SNP ID
rs79314587
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:173042913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTCTGGAGGAGAATGAAAGAGCG[A/T]TCACTAGAACAAAAGACAAAGCCAC
Phenotype
MIM: 603898
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TNFSF18 PubMed Links

Gene Details

Gene
TNFSF18
Gene Name
tumor necrosis factor superfamily member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005092.3 Intron NP_005083.2

View Full Product Details