Product Details

SNP ID
rs78779051
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:151009453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGCTCTCTGACTCAGTTGGACT[C/T]ACCCAGCCATGCTGAGCCGTCATGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM63A PubMed Links

Gene Details

Gene
FAM63A
Gene Name
family with sequence similarity 63 member A
There are no transcripts associated with this gene.

Gene
PRUNE1
Gene Name
prune exopolyphosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303229.1 Intron NP_001290158.1
NM_001303242.1 Intron NP_001290171.1
NM_001303243.1 Intron NP_001290172.1
NM_021222.2 Intron NP_067045.1
XM_005245393.4 Intron XP_005245450.1
XM_011509832.2 Intron XP_011508134.1
XM_017001955.1 Intron XP_016857444.1
XM_017001956.1 Intron XP_016857445.1
XM_017001957.1 Intron XP_016857446.1

View Full Product Details